missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TAF5 Partial ORF (NP_008882, 428 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006877-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16156215

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 14-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes an integral subunit of TFIID associated with all transcriptionally competent forms of that complex. This subunit interacts strongly with two TFIID subunits that show similarity to histones H3 and H4, and it may participate in forming a nucleosome-like core in the TFIID complex. [provided by RefSeq]

Sequence: PLPELKDSDKLDKIMNMKETTKRVRLGPDCLPSICFYTFLNAYQGLTAVDVTDDSSLIAGGFADSTVRVWSVTPKKLRSVKQASDLSLIDKESDDVLERIMDEKTASE
Especificaciones

Especificaciones

NP_008882
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PLPELKDSDKLDKIMNMKETTKRVRLGPDCLPSICFYTFLNAYQGLTAVDVTDDSSLIAGGFADSTVRVWSVTPKKLRSVKQASDLSLIDKESDDVLERIMDEKTASE
RUO
TAF5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6877
TAF5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TAF2D/TAFII100
TAF5
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales