missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SYK Partial ORF (AAH01645, 243 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006850-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: HYSYKADGLLRVLTVPCQKIGTQGNVNFGGRPQLPGSHPATWSAGGIISRIKSYSFPKPGHRKSSPAQGNRQESTVSFNPYEPELAPWAADKGPQREAEspecificaciones
AAH01645 | |
Liquid | |
6850 | |
SYK (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp313N1010/FLJ25043/FLJ37489 | |
SYK | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HYSYKADGLLRVLTVPCQKIGTQGNVNFGGRPQLPGSHPATWSAGGIISRIKSYSFPKPGHRKSSPAQGNRQESTVSFNPYEPELAPWAADKGPQREA | |
RUO | |
SYK | |
Wheat Germ (in vitro) | |
GST |