Learn More
Abnova™ Human SULT4A1 Partial ORF (NP_055166.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025830-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the sulfotransferase family. The encoded protein is a brain-specific sulfotransferase believed to be involved in the metabolism of neurotransmitters. Polymorphisms in this gene may be associated with susceptibility to schizophrenia. [provided by RefSeq]
Sequence: MAESEAETPSTPGEFESKYFEFHGVRLPPFCRGKMEEIANFPVRPSDVWIVTYPKSGTSLLQEVVYLVSQGADPDEIGLMNIDEQLPVLEYPQPGLDIIKEspecificaciones
NP_055166.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAESEAETPSTPGEFESKYFEFHGVRLPPFCRGKMEEIANFPVRPSDVWIVTYPKSGTSLLQEVVYLVSQGADPDEIGLMNIDEQLPVLEYPQPGLDIIK | |
RUO | |
SULT4A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25830 | |
SULT4A1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BR-STL-1/BRSTL1/DJ388M5.3/MGC40032/NST/SULTX3/hBR-STL-1 | |
SULT4A1 | |
Recombinant | |
wheat germ expression system |