Learn More
Abnova™ Human ST7L Full-length ORF (NP_060214.2, 1 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054879-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene was identified by its similarity to the ST7 tumor suppressor gene found in the chromosome 7q31 region. This gene is clustered in a tail-to-tail manner with the WNT2B gene in a chromosomal region known to be deleted and rearranged in a variety of cancers. The function of this gene has not yet been determined. Several transcript variants encoding many different isoforms have been described, but some have not been fully characterized. [provided by RefSeq]
Sequence: MADRGGVGEAAAVGASPASVPGLNPTLGWRERLRAGLAGTGASLWFVAGLGLLYALRIPLRLCENLAAVTVFLNSLTPKFYVALTGTSSLISGLIFIFEWWYFHKHGTSFIEQVSVSHLQPLMGGTESSISEPGSPSRNRENETSRQNLSECKVWRNPLNLFRGAEYRRYTWVTGKEPLTYYDMNLSAQDHQTFFTCDTDFLRPSDTVMQKAWRERNPPARIKAAYQALELNNDCATAYVLLAEEEATTIVDAERLFKQALKAGETIYRQSQQCQHQSPQHEAQLRRDTNVLVYIKRRLAMCARKLGRIREAVKIMRDLMKEFPPLTMLNIHENLLESLLELQAYPDVQAVLAKYDDISLPKSAAICYTAALLKTRTVSEKFSPETASRRGLSTAEINAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLQHWKRIEGALNLLQCTWEGTFRMIPYPLEKGHLFYPYPSCTETADRELLPTFHHVSVYPKKELPLFIHFTAGFCSSTAMIAILTHQFPEIMGIFAKAVLGLWCPQPWASSGFEENTQDLKSEDLGLSSGEspecificaciones
NP_060214.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FAM4B/FLJ11657/FLJ20284/ST7R/STLR | |
ST7L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54879 | |
ST7L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADRGGVGEAAAVGASPASVPGLNPTLGWRERLRAGLAGTGASLWFVAGLGLLYALRIPLRLCENLAAVTVFLNSLTPKFYVALTGTSSLISGLIFIFEWWYFHKHGTSFIEQVSVSHLQPLMGGTESSISEPGSPSRNRENETSRQNLSECKVWRNPLNLFRGAEYRRYTWVTGKEPLTYYDMNLSAQDHQTFFTCDTDFLRPSDTVMQKAWRERNPPARIKAAYQALELNNDCATAYVLLAEEEATTIVDAERLFKQALKAGETIYRQSQQCQHQSPQHEAQLRRDTNVLVYIKRRLAMCARKLGRIREAVKIMRDLMKEFPPLTMLNIHENLLESLLELQAYPDVQAVLAKYDDISLPKSAAICYTAALLKTRTVSEKFSPETASRRGLSTAEINAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLQHWKRIEGALNLLQCTWEGTFRMIPYPLEKGHLFYPYPSCTETADRELLPTFHHVSVYPKKELPLFIHFTAGFCSSTAMIAILTHQFPEIMGIFAKAVLGLWCPQPWASSGFEENTQDLKSEDLGLSSG | |
RUO | |
ST7L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |