missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC4A8 Partial ORF (NP_004849, 187 a.a. - 286 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009498-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: IEEISDLILDQQELSSDLNDSMRVKVREALLKKHHHQNEKKRNNLIPIVRSFAEVGKKQSDPHLMDKHGQTVSPQSVPTTNLEVKNGVNCEHSPVDLSKVEspecificaciones
NP_004849 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IEEISDLILDQQELSSDLNDSMRVKVREALLKKHHHQNEKKRNNLIPIVRSFAEVGKKQSDPHLMDKHGQTVSPQSVPTTNLEVKNGVNCEHSPVDLSKV | |
RUO | |
SLC4A8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9498 | |
SLC4A8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761B2318/FLJ46462/NBC3 | |
SLC4A8 | |
Recombinant | |
wheat germ expression system |