Learn More
Abnova™ Human SLC3A1 Partial ORF (AAH22386, 211 a.a. - 320 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006519-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract. Mutations and deletions in this gene are associated with cystinuria. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]
Sequence: FIPNHTSDKHIWFQLSRTRTGKYTDYYIWHDCTHENGKTIPPNNWLSVYGNSSWHFDEVRNQCYFHQFMKEQPDLNFRNPDVQEEIKEILRFWLTKGVDGFSLDAVKFLLEspecificaciones
AAH22386 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FIPNHTSDKHIWFQLSRTRTGKYTDYYIWHDCTHENGKTIPPNNWLSVYGNSSWHFDEVRNQCYFHQFMKEQPDLNFRNPDVQEEIKEILRFWLTKGVDGFSLDAVKFLL | |
RUO | |
SLC3A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6519 | |
SLC3A1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATR1/CSNU1/D2H/FLJ34681/NBAT/RBAT | |
SLC3A1 | |
Recombinant | |
wheat germ expression system |