missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC35F2 Full-length ORF (NP_059985.2, 1 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054733-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MEADSPAGPGAPEPLAEGAAAEFSSLLRRIKGKLFTWNILKTIALGQMLSLCICGTAITSQYLAERYKVNTPMLQSFINYCLLFLIYTVMLAFRSGSDNLLVILKRKWWKYILLGLADVEANYVIVRAYQYTTLTSVQLLDCFGIPVLMALSWFILHARYRVIHFIAVAVCLLGVGTMVGADILAGREDNSGSDVLIGDILVLLGASLYAISNVCEEYIVKKLSRQEFLGMVGLFGTIISGIQLLIVEYKDIASIHWDWKIALLFVAFALCMFCLYSFMPLVIKVTSATSVNLGILTADLYSLFVGLFLFGYKFSGLYILSFTVIMVGFILYCSTPTRTAEPAESSVPPVTSIGIDNLGLKLEENLQETHSAVLEspecificaciones
NP_059985.2 | |
Liquid | |
54733 | |
SLC35F2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEADSPAGPGAPEPLAEGAAAEFSSLLRRIKGKLFTWNILKTIALGQMLSLCICGTAITSQYLAERYKVNTPMLQSFINYCLLFLIYTVMLAFRSGSDNLLVILKRKWWKYILLGLADVEANYVIVRAYQYTTLTSVQLLDCFGIPVLMALSWFILHARYRVIHFIAVAVCLLGVGTMVGADILAGREDNSGSDVLIGDILVLLGASLYAISNVCEEYIVKKLSRQEFLGMVGLFGTIISGIQLLIVEYKDIASIHWDWKIALLFVAFALCMFCLYSFMPLVIKVTSATSVNLGILTADLYSLFVGLFLFGYKFSGLYILSFTVIMVGFILYCSTPTRTAEPAESSVPPVTSIGIDNLGLKLEENLQETHSAVL | |
RUO | |
SLC35F2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp667H1615/FLJ13018/HSNOV1 | |
SLC35F2 | |
Yes | |
wheat germ expression system |