Learn More
Abnova™ Human SLC26A4 Partial ORF (NP_000432, 674 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005172-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Mutations in this gene are associated with Pendred syndrome, the most common form of syndromic deafness, an autosomal-recessive disease. It is highly homologous to the SLC26A3 gene; they have similar genomic structures and this gene is located 3' of the SLC26A3 gene. The encoded protein has homology to sulfate transporters. [provided by RefSeq]
Sequence: RSLRVIVKEFQRIDVNVYFASLQDYVIEKLEQCGFFDDNIRKDTFFLTVHDAILYLQNQVKSQEGQGSILETITLIQDCKDEspecificaciones
NP_000432 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.65kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RSLRVIVKEFQRIDVNVYFASLQDYVIEKLEQCGFFDDNIRKDTFFLTVHDAILYLQNQVKSQEGQGSILETITLIQDCKD | |
RUO | |
SLC26A4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5172 | |
SLC26A4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB4/EVA/PDS | |
SLC26A4 | |
Recombinant | |
wheat germ expression system |