Learn More
Abnova™ Human SIPA1 Partial ORF (AAH10492, 933 a.a. - 1042 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006494-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date. [provided by RefSeq]
Sequence: SEIASTCNTILESLSREGQPIPESGDPKGTPKSDAEPEPGNLSEKVSHLESMLRKLQEDLQKEKADRAALEEEVRSLRHNNRRLQAESESAATRLLLASKQLGSPTADLAEspecificaciones
AAH10492 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEIASTCNTILESLSREGQPIPESGDPKGTPKSDAEPEPGNLSEKVSHLESMLRKLQEDLQKEKADRAALEEEVRSLRHNNRRLQAESESAATRLLLASKQLGSPTADLA | |
RUO | |
SIPA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6494 | |
SIPA1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC102688/MGC17037/SPA1 | |
SIPA1 | |
Recombinant | |
wheat germ expression system |