missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SH3PX3 Full-length ORF (NP_695003.1, 1 a.a. - 574 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00257364-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MALKGRALYDFHSENKEEISIQQDEDLVIFSETSLDGWLQGQNSRGETGLFPASYVEIVRSGISTNHADYSSSPAGSPGAQVSLYNSPSVASPARSGGGSGFLSNQGSFEEDDDDDWDDWDDGCTVVEEPRAGGLGTNGHPPLNLSYPGAYPSQHMAFRPKPPLERQDSLASAKRGSVVGRNLNRFSCFVRSGVEAFILGDVPMMAKIAETYSIEMGPRGPQWKANPHPFACSVEDPTKQTKFKGIKSYISYKLTPTHAASPVYRRYKHFDWLYNRLLHKFTVISVPHLPEKQATGRFEEDFIEKRKRRLILWMDHMTSHPVLSQYEGFQHFLSCLDDKQWKMGKRRAEKDEMVGASFLLTFQIPTEHQDLQDVEDRVDTFKAFSKKMDDSVLQLSTVASELVRKHVGGFRKEFQKLGSAFQAISHSFQMDPPFCSEALNSAISHTGRTYEAIGEMFAEQPKNDLFQMLDTLSLYQGLLSNFPDIIHLQKGAFAKVKESQRMSDEGRMVQDEADGIRRRCRVVGFALQAEMNHFHQRRELDFKHMMQNYLRQQILFYQRVGQQLEKTLRMYDNLEspecificaciones
NP_695003.1 | |
Liquid | |
257364 | |
SH3PX3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALKGRALYDFHSENKEEISIQQDEDLVIFSETSLDGWLQGQNSRGETGLFPASYVEIVRSGISTNHADYSSSPAGSPGAQVSLYNSPSVASPARSGGGSGFLSNQGSFEEDDDDDWDDWDDGCTVVEEPRAGGLGTNGHPPLNLSYPGAYPSQHMAFRPKPPLERQDSLASAKRGSVVGRNLNRFSCFVRSGVEAFILGDVPMMAKIAETYSIEMGPRGPQWKANPHPFACSVEDPTKQTKFKGIKSYISYKLTPTHAASPVYRRYKHFDWLYNRLLHKFTVISVPHLPEKQATGRFEEDFIEKRKRRLILWMDHMTSHPVLSQYEGFQHFLSCLDDKQWKMGKRRAEKDEMVGASFLLTFQIPTEHQDLQDVEDRVDTFKAFSKKMDDSVLQLSTVASELVRKHVGGFRKEFQKLGSAFQAISHSFQMDPPFCSEALNSAISHTGRTYEAIGEMFAEQPKNDLFQMLDTLSLYQGLLSNFPDIIHLQKGAFAKVKESQRMSDEGRMVQDEADGIRRRCRVVGFALQAEMNHFHQRRELDFKHMMQNYLRQQILFYQRVGQQLEKTLRMYDNL | |
RUO | |
SNX33 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC32065/SH3PX3/SH3PXD3C/SNX30 | |
SNX33 | |
Yes | |
wheat germ expression system |