missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SGSM3 Full-length ORF (CAK54470.1, 1 a.a. - 749 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027352-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MSGSHTPACGPFSALTPSIWPQEILAKYTQKEESAEQPEFYYDEFGFRVYKEEGDEPGSSLLANSPLMEDAPQRLRWQAHLEFTHNHDVGDLTWDKIAVSLPRSEKLRSLVLAGIPHGMRPQLWMRLSGALQKKRNSELSYREIVKNSSNDETIAAKQIEKDLLRTMPSNACFASMGSIGVPRLRRVLRALAWLYPEIGYCQGTGMVAACLLLFLEEEDAFWMMSAIIEDLLPASYFSTTLLGVQTDQRVLRHLIVQYLPRLDKLLQEHDIELSLITLHWFLTAFASVVDIKLLLRIWDLFFYEGSRVLFQLTLGMLHLKEEELIQSENSASIFNTLSDIPSQMEDAELLLGVAMRLAGSLTDVAVETQRRKHLAYLIADQGQLLGAGTLTNLSQVVRRRTQRRKSTITALLFGEDDLEALKAKNIKQTELVADLREAILRVARHFQCTDPKNCSVELTPDYSMESHQRDHENYVACSRSHRRRAKALLDFERHDDDELGFRKNDIITIVSQKDEHCWVGELNGLRGWFPAKFVEVLDERSKEYSIAGDDSVTEGVTDLVRGTLCPALKALFEHGLKKPSLLGGACHPWLFIEEAAGREVERDFASVYSRLVLCKTFRLDEDGKVLTPEELLYRAVQSVNVTHDAVHAQMDVKLRSLICVGLNEQVLHLWLEVLCSSLPTVEKWYQPWSFLRSPGWVQIKCELRVLCCFAFSLSQDWELPAKREAQQPLKEGVRDMLVKHHLFSWDVDGEspecificaciones
CAK54470.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
108.79kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761D051/MAP/RUSC3/RUTBC3 | |
SGSM3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27352 | |
SGSM3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSGSHTPACGPFSALTPSIWPQEILAKYTQKEESAEQPEFYYDEFGFRVYKEEGDEPGSSLLANSPLMEDAPQRLRWQAHLEFTHNHDVGDLTWDKIAVSLPRSEKLRSLVLAGIPHGMRPQLWMRLSGALQKKRNSELSYREIVKNSSNDETIAAKQIEKDLLRTMPSNACFASMGSIGVPRLRRVLRALAWLYPEIGYCQGTGMVAACLLLFLEEEDAFWMMSAIIEDLLPASYFSTTLLGVQTDQRVLRHLIVQYLPRLDKLLQEHDIELSLITLHWFLTAFASVVDIKLLLRIWDLFFYEGSRVLFQLTLGMLHLKEEELIQSENSASIFNTLSDIPSQMEDAELLLGVAMRLAGSLTDVAVETQRRKHLAYLIADQGQLLGAGTLTNLSQVVRRRTQRRKSTITALLFGEDDLEALKAKNIKQTELVADLREAILRVARHFQCTDPKNCSVELTPDYSMESHQRDHENYVACSRSHRRRAKALLDFERHDDDELGFRKNDIITIVSQKDEHCWVGELNGLRGWFPAKFVEVLDERSKEYSIAGDDSVTEGVTDLVRGTLCPALKALFEHGLKKPSLLGGACHPWLFIEEAAGREVERDFASVYSRLVLCKTFRLDEDGKVLTPEELLYRAVQSVNVTHDAVHAQMDVKLRSLICVGLNEQVLHLWLEVLCSSLPTVEKWYQPWSFLRSPGWVQIKCELRVLCCFAFSLSQDWELPAKREAQQPLKEGVRDMLVKHHLFSWDVDG | |
RUO | |
SGSM3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |