missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEMA3A Partial ORF (NP_006071, 672 a.a. - 770 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010371-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16164952

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type (immunoglobulin-like) domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease. [provided by RefSeq]

Sequence: HLEELLHKDDDGDGSKTKEMSNSMTPSQKVWYRDFMQLINHPNLNTMDEFCEQVWKRDRKQRRQRPGHTPGNSNKWKHLQENKKGRNRRTHEFERAPRS
Especificaciones

Especificaciones

NP_006071
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HLEELLHKDDDGDGSKTKEMSNSMTPSQKVWYRDFMQLINHPNLNTMDEFCEQVWKRDRKQRRQRPGHTPGNSNKWKHLQENKKGRNRRTHEFERAPRS
RUO
SEMA3A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10371
SEMA3A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Hsema-I/Hsema-III/MGC133243/SEMA1/SEMAD/SEMAIII/SEMAL/SemD/coll-1
SEMA3A
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales