Learn More
Abnova™ Human RTDR1 Partial ORF (NP_055248.1, 249 a.a. - 348 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027156-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with no known function but with slight similarity to a yeast vacuolar protein. The gene is located in a region deleted in pediatric rhabdoid tumors of the brain, kidney and soft tissues, but mutations in this gene have not been associated with the disease. [provided by RefSeq]
Sequence: SNAAGALMFATVITEGKYAALEAQAIGLLLELLHSPMTIARLNATKALTMLAEAPEGRKALQTHVPTFRAMEVETYEKPQVAEALQRAARIAISVIEFKPEspecificaciones
NP_055248.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SNAAGALMFATVITEGKYAALEAQAIGLLLELLHSPMTIARLNATKALTMLAEAPEGRKALQTHVPTFRAMEVETYEKPQVAEALQRAARIAISVIEFKP | |
RUO | |
RTDR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27156 | |
RTDR1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC16968 | |
RTDR1 | |
Recombinant | |
wheat germ expression system |