Learn More
Abnova™ Human RPL12 Partial ORF (NP_000967, 66 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006136-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L11P family of ribosomal proteins. It is located in the cytoplasm. The protein binds directly to the 26S rRNA. This gene is co-transcribed with the U65 snoRNA, which is located in its fourth intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]
Sequence: NRQAQIEVVPSASALIIKALKEPPRDRKKQKNIKHSGNITFDEIVNIARQMRHRSLARELSGTIKEILGTAQSVGCNVDGRHPHDIIDDINSGAVECPASEspecificaciones
NP_000967 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NRQAQIEVVPSASALIIKALKEPPRDRKKQKNIKHSGNITFDEIVNIARQMRHRSLARELSGTIKEILGTAQSVGCNVDGRHPHDIIDDINSGAVECPAS | |
RUO | |
RPL12 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6136 | |
RPL12 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPL12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |