Learn More
Abnova™ Human RNF32 Full-length ORF (AAH15416, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00140545-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene contains two RING ring finger motifs. RING finger motifs are present in a variety of functionally distinct proteins and are known to be involved in protein-DNA or protein-protein interactions. This gene was found to be expressed during spermatogenesis, most likely in spermatocytes and/or in spermatids. Several alternatively spliced transcript variants exist, but their full length natures are not clear. [provided by RefSeq]
Sequence: MLKNKGHSSKKDNLAVNAVALQDHILHDLQLRNLSVADHSKTQVQKKENKSLKRDTKAIIDTGLKKTTQCPKLEDSEKEYVLDPKPPPLTLAQKLGLIGPPPPPLSSDEWEKVKQRSLLQGDSVQPCPICKEEFELRPQVLLSCSHVFHKACLQAFEKFTNKKTCPLCRKNQYQTRVIHDGARLFRIKCVTRIQAYWRGCVVRKWYRNLRKTVPPTDAKLRKKFFEKKTQDWKPAEspecificaciones
AAH15416 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.48kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKSG33/HSD15 | |
RNF32 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
140545 | |
RNF32 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLKNKGHSSKKDNLAVNAVALQDHILHDLQLRNLSVADHSKTQVQKKENKSLKRDTKAIIDTGLKKTTQCPKLEDSEKEYVLDPKPPPLTLAQKLGLIGPPPPPLSSDEWEKVKQRSLLQGDSVQPCPICKEEFELRPQVLLSCSHVFHKACLQAFEKFTNKKTCPLCRKNQYQTRVIHDGARLFRIKCVTRIQAYWRGCVVRKWYRNLRKTVPPTDAKLRKKFFEKKTQDWKPA | |
RUO | |
RNF32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |