missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF32 Full-length ORF (AAH15416, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00140545-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108203

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene contains two RING ring finger motifs. RING finger motifs are present in a variety of functionally distinct proteins and are known to be involved in protein-DNA or protein-protein interactions. This gene was found to be expressed during spermatogenesis, most likely in spermatocytes and/or in spermatids. Several alternatively spliced transcript variants exist, but their full length natures are not clear. [provided by RefSeq]

Sequence: MLKNKGHSSKKDNLAVNAVALQDHILHDLQLRNLSVADHSKTQVQKKENKSLKRDTKAIIDTGLKKTTQCPKLEDSEKEYVLDPKPPPLTLAQKLGLIGPPPPPLSSDEWEKVKQRSLLQGDSVQPCPICKEEFELRPQVLLSCSHVFHKACLQAFEKFTNKKTCPLCRKNQYQTRVIHDGARLFRIKCVTRIQAYWRGCVVRKWYRNLRKTVPPTDAKLRKKFFEKKTQDWKPA
Especificaciones

Especificaciones

AAH15416
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.48kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FKSG33/HSD15
RNF32
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
140545
RNF32 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLKNKGHSSKKDNLAVNAVALQDHILHDLQLRNLSVADHSKTQVQKKENKSLKRDTKAIIDTGLKKTTQCPKLEDSEKEYVLDPKPPPLTLAQKLGLIGPPPPPLSSDEWEKVKQRSLLQGDSVQPCPICKEEFELRPQVLLSCSHVFHKACLQAFEKFTNKKTCPLCRKNQYQTRVIHDGARLFRIKCVTRIQAYWRGCVVRKWYRNLRKTVPPTDAKLRKKFFEKKTQDWKPA
RUO
RNF32
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales