Learn More
Abnova™ Human RNF121 Partial ORF (NP_060790.2, 193 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055298-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Several alternatively spliced transcript variants have been noted for this gene, however, not all are likely to encode viable protein products. [provided by RefSeq]
Sequence: ERDFAEMCADYMASTIGFYSESGMPTKHLSDSVCAVCGQQIFVDVSEEGIIENTYRLSCNHVFHEFCIRGWCIVGKKQTCPYCKEKVDLKRMFSNPWERPHVMYGQLLDEspecificaciones
NP_060790.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ERDFAEMCADYMASTIGFYSESGMPTKHLSDSVCAVCGQQIFVDVSEEGIIENTYRLSCNHVFHEFCIRGWCIVGKKQTCPYCKEKVDLKRMFSNPWERPHVMYGQLLD | |
RUO | |
RNF121 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55298 | |
RNF121 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11099 | |
RNF121 | |
Recombinant | |
wheat germ expression system |