missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNASE2 Partial ORF (NP_002925, 86 a.a. - 161 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006036-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: NMTCPSNKTRKNCHHSGSQVPLIHCNLTTPSPQNISNCRYAQTPANMFYIVACDNRDQRRDPPQYPVVPVHLDRIIEspecificaciones
NP_002925 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NMTCPSNKTRKNCHHSGSQVPLIHCNLTTPSPQNISNCRYAQTPANMFYIVACDNRDQRRDPPQYPVVPVHLDRII | |
RUO | |
RNASE2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6036 | |
RNASE2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EDN/RNS2 | |
RNASE2 | |
Recombinant | |
wheat germ expression system |