missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RIN1 Partial ORF (AAH14417, 646 a.a. - 755 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009610-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: VPPEASIATLNQLCATKFRVTQPNTFGLFLYKEQGYHRLPPGALAHRLPTTGYLVYRRAEWPETQGAVTEEEGSGQSEARSRGEEQGCQGDGDAGVKASPRDIREQSETTEspecificaciones
AAH14417 | |
Liquid | |
9610 | |
RIN1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RIN1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VPPEASIATLNQLCATKFRVTQPNTFGLFLYKEQGYHRLPPGALAHRLPTTGYLVYRRAEWPETQGAVTEEEGSGQSEARSRGEEQGCQGDGDAGVKASPRDIREQSETT | |
RUO | |
RIN1 | |
Yes | |
wheat germ expression system |