Learn More
Abnova™ Human RFX1 Partial ORF (NP_002909, 502 a.a. - 600 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005989-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X2, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with RFX family members X2, X3, and X5, but not with X4. This protein binds to the X-boxes of MHC class II genes and is essential for their expression. Also, it can bind to an inverted repeat that is required for expression of hepatitis B virus genes. [provided by RefSeq]
Sequence: SKYHYYGLRIKASSPLLRLMEDQQHMAMRGQPFSQKQRLKPIQKMEGMTNGVAVGQQPSTGLSDISAQVQQYQQFLDASRSLPDFTELDLQGKVLPEGVEspecificaciones
NP_002909 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SKYHYYGLRIKASSPLLRLMEDQQHMAMRGQPFSQKQRLKPIQKMEGMTNGVAVGQQPSTGLSDISAQVQQYQQFLDASRSLPDFTELDLQGKVLPEGV | |
RUO | |
RFX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5989 | |
RFX1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EF-C | |
RFX1 | |
Recombinant | |
wheat germ expression system |