missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RFP2 Full-length ORF (NP_001007279.1, 1 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010206-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16172106

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This gene is located on chromosome 13 within the minimal deletion region for B-cell chronic lymphocytic leukemia. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]

Sequence: MDVMELLEEDLTCPICCSLFDDPRVLPCSHNFCKKCLEGILEGSVRNSLWRPAPFKCPTCRKETSATGINSLQVNYSLKGIVEKYNKIKISPKMPVCKGHLGQPLNIFCLTDMQLICGICATRGEHTKHVFCSIEDAYAQERDAFESLFQSFETWRRGDALSRLDTLETSKRKSLQLLTKDSDKVKEFFEKLQHTLDQKKNEILSDFETMKLAVMQAYDPEINKLNTILQEQRMAFNIAEAFKDVSEPIVFLQQMQEFREKIKVIKETPLPPSNLPASPLMKNFDTSQWEDIKLVDVDKLSLPQDTGTFISKIPWSFYKLFLLILLLGLVIVFGPTMFLEWSLFDDLATWKGCLSNFSSYLTKTADFIEQSVFYWEQVTDGFFIFNERFKNFTLVVLNNVAEFVCKYKLL
Especificaciones

Especificaciones

NP_001007279.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAR/DLEU5/LEU5/RFP2/RNF77
TRIM13
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10206
RFP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDVMELLEEDLTCPICCSLFDDPRVLPCSHNFCKKCLEGILEGSVRNSLWRPAPFKCPTCRKETSATGINSLQVNYSLKGIVEKYNKIKISPKMPVCKGHLGQPLNIFCLTDMQLICGICATRGEHTKHVFCSIEDAYAQERDAFESLFQSFETWRRGDALSRLDTLETSKRKSLQLLTKDSDKVKEFFEKLQHTLDQKKNEILSDFETMKLAVMQAYDPEINKLNTILQEQRMAFNIAEAFKDVSEPIVFLQQMQEFREKIKVIKETPLPPSNLPASPLMKNFDTSQWEDIKLVDVDKLSLPQDTGTFISKIPWSFYKLFLLILLLGLVIVFGPTMFLEWSLFDDLATWKGCLSNFSSYLTKTADFIEQSVFYWEQVTDGFFIFNERFKNFTLVVLNNVAEFVCKYKLL
RUO
TRIM13
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales