missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RFC3 Partial ORF (AAH00149, 257 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005983-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16124295

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 38 kDa subunit. This subunit is essential for the interaction between the 140 kDa subunit and the core complex that consists of the 36, 37, and 40 kDa subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]

Sequence: ETANAIVSQQTPQRLLEVRGRLYELLTHCIPPEIIMKGLLSELLHNCDGQLKGEVAQMAAYYEHRLQLGSKAIYHLEAFVAKFMALYKKFMEDGLEGMMF
Especificaciones

Especificaciones

AAH00149
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ETANAIVSQQTPQRLLEVRGRLYELLTHCIPPEIIMKGLLSELLHNCDGQLKGEVAQMAAYYEHRLQLGSKAIYHLEAFVAKFMALYKKFMEDGLEGMMF
RUO
RFC3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5983
RFC3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC5276/RFC38
RFC3
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales