Learn More
STEMCELL Technologies Human Recombinant G-CSF, ACF
Granulocyte colony-stimulating factor, animal component-free
Marca:  STEMCELL Technologies 78138
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17199651
  • 266.00€ / 10 microgramos
Fecha estimada de envío: 04-04-2024
para ver el stock
Descripción
Descripción
  • Granulocyte colony-stimulating factor (G-CSF) is a member of the CSF family of glycoproteins that regulate hematopoietic cell proliferation, differentiation, and function
  • It is a key cytokine involved in the production of neutrophils and the stimulation of granulocyte colony formation from hematopoietic progenitor cells
  • G-CSF causes a range of effects including a transient reduction of SDF-1 expression, the activation of metalloproteases that cleave VCAM-1, and the release of norepinephrine from the sympathetic nervous system, leading to the release or mobilization of hematopoietic stem cells from the bone marrow into the periphery
  • The G-CSF receptor is expressed on a variety of hematopoietic cells, including myeloid-committed progenitor cells, neutrophils, granulocytes, and monocytes
Especificaciones
Especificaciones
Colony-stimulating factor 3, CSF-3, MGI-1G, Pluripoietin
10 μg
Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neurons
≥95%