Learn More
STEMCELL Technologies Human Recombinant BMP-4
Bone morphogenetic protein 4
Marca:  STEMCELL Technologies 78211
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17199841
  • 512.05€ / 20 microgramos
Fecha estimada de envío: 04-04-2024
para ver el stock
Descripción
Descripción
  • Bone morphogenetic protein 4 (BMP-4) is a member of the highly conserved transforming growth factor β (TGF-β) superfamily
  • Mature BMP-4 is a disulfide-linked homodimeric protein consisting of two 116-amino-acid residue subunits, and is generated by the proteolytic removal of the signal peptide and propeptides (Xiao et al.) BMP-4 binds to type I and type II receptors on cells
  • This binding results in the phosphorylation of receptor 1, which in turn results in the phosphorylation of Smad proteins, which then go on to act as transcription factors (Zhang et al.) BMPs have been shown to be key regulators of embryogenesis and are known to play a role in the growth and differentiation of various cell types, including embryonic stem (ES) cells, induced pluripotent stem (iPS) cells, mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells (Chadwick et al.; Graham et al.; Jones et al.; Lengerke et al.; Zhang et al.)
Especificaciones
Especificaciones
BMP-2B, BMP2B1, MCOPS6, OFC11, ZYME
20 μg
Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells
≥95%