Learn More
Abnova™ Human RCBTB1 Partial ORF (NP_060661.3, 2 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055213-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with an N-terminal RCC1 domain and a C-terminal BTB (broad complex, tramtrack and bric-a-brac) domain. In rat, over-expression of this gene in vascular smooth muscle cells induced cellular hypertrophy. In rat, the C-terminus of RCBTB1 interacts with the angiotensin II receptor-1A. In humans, this gene maps to a region of chromosome 13q that is frequently deleted in B-cell chronic lymphocytic leukemia and other lymphoid malignancies. [provided by RefSeq]
Sequence: VDVGKWPIFTLLSPQEIASIRKACVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLSEspecificaciones
NP_060661.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VDVGKWPIFTLLSPQEIASIRKACVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLS | |
RUO | |
RCBTB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55213 | |
RCBTB1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLLD7/CLLL7/GLP/MGC33184/RP11-185C18.1 | |
RCBTB1 | |
Recombinant | |
wheat germ expression system |