Learn More
Abnova™ Human RBM12 Full-length ORF (NP_006038.2, 1 a.a. - 932 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010137-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5' UTR results in two transcript variants. Both variants encode the same protein. [provided by RefSeq]
Sequence: MAVVIRLQGLPIVAGTMDIRHFFSGLTIPDGGVHIVGGELGEAFIVFATDEDARLGMMRTGGTIKGSKVTLLLSSKTEMQNMIELSRRRFETANLDIPPANASRSGPPPSSGMSSRVNLPTTVSNFNNPSPSVVTATTSVHESNKNIQTFSTASVGTAPPNMGASFGSPTFSSTVPSTASPMNTVPPPPIPPIPAMPSLPPMPSIPPIPVPPPVPTLPPVPPVPPIPPVPSVPPMTPLPPMSGMPPLNPPPVAPLPAGMNGSGAPMNLNNNLNPMFLGPLNPVNPIQMNSQSSVKPLPINPDDLYVSVHGMPFSAMENDVRDFFHGLRVDAVHLLKDHVGRNNGNGLVKFLSPQDTFEALKRNRMLMIQRYVEVSPATERQWVAAGGHITFKQNMGPSGQTHPPPQTLPRSKSPSGQKRSRSRSPHEAGFCVYLKGLPFEAENKHVIDFFKKLDIVEDSIYIAYGPNGKATGEGFVEFRNEADYKAALCRHKQYMGNRFIQVHPITKKGMLEKIDMIRKRLQNFSYDQREMILNPEGDVNSAKVCAHITNIPFSITKMDVLQFLEGIPVDENAVHVLVDNNGQGLGQALVQFKNEDDARKSERLHRKKLNGREAFVHVVTLEDMREIEKNPPAQGKKGLKMPVPGNPAVPGMPNAGLPGVGLPSAGLPGAGLPSTGLPGSAITSAGLPGAGMPSAGIPSAGGEEHAFLTVGSKEANNGPPFNFPGNFGGSNAFGPPIPPPGLGGGAFGDARPGMPSVGNSGLPGLGLDVPGFGGGPNNLSGPSGFGGGPQNFGNGPGSLGGPPGFGSGPPGLGSAPGHLGGPPAFGPGPGPGPGPGPIHIGGPPGFASSSGKPGPTVIKVQNMPFTVSIDEILDFFYGYQVIPGSVCLKYNEKGMPTGEAMVAFESRDEATAAVIDLNDRPIGSRKVKLVLGEspecificaciones
NP_006038.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
123.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HRIHFB2091/KIAA0765/SWAN | |
RBM12 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10137 | |
RBM12 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVVIRLQGLPIVAGTMDIRHFFSGLTIPDGGVHIVGGELGEAFIVFATDEDARLGMMRTGGTIKGSKVTLLLSSKTEMQNMIELSRRRFETANLDIPPANASRSGPPPSSGMSSRVNLPTTVSNFNNPSPSVVTATTSVHESNKNIQTFSTASVGTAPPNMGASFGSPTFSSTVPSTASPMNTVPPPPIPPIPAMPSLPPMPSIPPIPVPPPVPTLPPVPPVPPIPPVPSVPPMTPLPPMSGMPPLNPPPVAPLPAGMNGSGAPMNLNNNLNPMFLGPLNPVNPIQMNSQSSVKPLPINPDDLYVSVHGMPFSAMENDVRDFFHGLRVDAVHLLKDHVGRNNGNGLVKFLSPQDTFEALKRNRMLMIQRYVEVSPATERQWVAAGGHITFKQNMGPSGQTHPPPQTLPRSKSPSGQKRSRSRSPHEAGFCVYLKGLPFEAENKHVIDFFKKLDIVEDSIYIAYGPNGKATGEGFVEFRNEADYKAALCRHKQYMGNRFIQVHPITKKGMLEKIDMIRKRLQNFSYDQREMILNPEGDVNSAKVCAHITNIPFSITKMDVLQFLEGIPVDENAVHVLVDNNGQGLGQALVQFKNEDDARKSERLHRKKLNGREAFVHVVTLEDMREIEKNPPAQGKKGLKMPVPGNPAVPGMPNAGLPGVGLPSAGLPGAGLPSTGLPGSAITSAGLPGAGMPSAGIPSAGGEEHAFLTVGSKEANNGPPFNFPGNFGGSNAFGPPIPPPGLGGGAFGDARPGMPSVGNSGLPGLGLDVPGFGGGPNNLSGPSGFGGGPQNFGNGPGSLGGPPGFGSGPPGLGSAPGHLGGPPAFGPGPGPGPGPGPIHIGGPPGFASSSGKPGPTVIKVQNMPFTVSIDEILDFFYGYQVIPGSVCLKYNEKGMPTGEAMVAFESRDEATAAVIDLNDRPIGSRKVKLVLG | |
RUO | |
RBM12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |