Learn More
Abnova™ Human RASSF3 Full-length ORF (NP_835463.1, 1 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00283349-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The RAS oncogene (MIM 190020) is mutated in nearly one-third of all human cancers. Members of the RAS superfamily are plasma membrane GTP-binding proteins that modulate intracellular signal transduction pathways. A subfamily of RAS effectors, including RASSF3, share a RAS association (RA) domain.[supplied by OMIM]
Sequence: MSSGYSSLEEDAEDFFFTARTSFFRRAPQGKPRSGQQDVEKEKETHSYLSKEEIKEKVHKYNLAVTDKLKMTLNSNGIYTGFIKVQMELCKPPQTSPNSGKLSPSSNGCMNTLHISSTNTVGEVIEALLKKFLVTESPAKFALYKRCHREDQVYACKLSDREHPLYLRLVAGPRTDTLSFVLREHEIGEWEAFSLPELQNFLRILDKEEDEQLQNLKRRYTAYRQKLEEALREVWKPDEspecificaciones
NP_835463.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119194/MGC119195/MGC119197/RASSF5 | |
RASSF3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
283349 | |
RASSF3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSGYSSLEEDAEDFFFTARTSFFRRAPQGKPRSGQQDVEKEKETHSYLSKEEIKEKVHKYNLAVTDKLKMTLNSNGIYTGFIKVQMELCKPPQTSPNSGKLSPSSNGCMNTLHISSTNTVGEVIEALLKKFLVTESPAKFALYKRCHREDQVYACKLSDREHPLYLRLVAGPRTDTLSFVLREHEIGEWEAFSLPELQNFLRILDKEEDEQLQNLKRRYTAYRQKLEEALREVWKPD | |
RUO | |
RASSF3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |