Learn More
Abnova™ Human RASSF2 Full-length ORF (NP_055552.1, 1 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009770-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that contains a Ras association domain. Similar to its cattle and sheep counterparts, this gene is located near the prion gene. Two alternatively spliced transcripts encoding the same isoform have been reported. [provided by RefSeq]
Sequence: MDYSHQTSLVPCGQDKYISKNELLLHLKTYNLYYEGQNLQLRHREEEDEFIVEGLLNISWGLRRPIRLQMQDDNERIRPPPSSSSWHSGCNLGAQGTTLKPLTVPKVQISEVDAPPEGDQMPSSTDSRGLKPLQEDTPQLMRTRSDVGVRRRGNVRTPSDQRRIRRHRFSINGHFYNHKTSVFTPAYGSVTNVRINSTMTTPQVLKLLLNKFKIENSAEEFALYVVHTSGEKQKLKATDYPLIARILQGPCEQISKVFLMEKDQVEEVTYDVAQYIKFEMPVLKSFIQKLQEEEDREVKKLMRKYTVLRLMIRQRLEEIAETPATIEspecificaciones
NP_055552.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781O1747/KIAA0168/RASFADIN | |
RASSF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9770 | |
RASSF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDYSHQTSLVPCGQDKYISKNELLLHLKTYNLYYEGQNLQLRHREEEDEFIVEGLLNISWGLRRPIRLQMQDDNERIRPPPSSSSWHSGCNLGAQGTTLKPLTVPKVQISEVDAPPEGDQMPSSTDSRGLKPLQEDTPQLMRTRSDVGVRRRGNVRTPSDQRRIRRHRFSINGHFYNHKTSVFTPAYGSVTNVRINSTMTTPQVLKLLLNKFKIENSAEEFALYVVHTSGEKQKLKATDYPLIARILQGPCEQISKVFLMEKDQVEEVTYDVAQYIKFEMPVLKSFIQKLQEEEDREVKKLMRKYTVLRLMIRQRLEEIAETPATI | |
RUO | |
RASSF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |