Learn More
Abnova™ Human RANBP3 Partial ORF (AAH04349, 298 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008498-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with a RanBD1 domain that is found in both the nucleus and cytoplasm. This protein plays a role in nuclear export as part of a heteromeric complex. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: KESLAESAAAYTKATARKCLLEKVEVITGEEAESNVLQMQCKLFVFDKTSQSWLLRHPHPSHPAVGPCLCGEQPALCPCPGLPNYRPGTPAQLGGLLVRVEspecificaciones
AAH04349 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KESLAESAAAYTKATARKCLLEKVEVITGEEAESNVLQMQCKLFVFDKTSQSWLLRHPHPSHPAVGPCLCGEQPALCPCPGLPNYRPGTPAQLGGLLVRV | |
RUO | |
RANBP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8498 | |
RANBP3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586I1520 | |
RANBP3 | |
Recombinant | |
wheat germ expression system |