Learn More
Abnova™ Human RAD18 Full-length ORF (NP_064550.2, 1 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056852-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is highly similar to S. cerevisiae DNA damage repair protein Rad18. Yeast Rad18 functions through its interaction with Rad6, which is an ubiquitin-conjugating enzyme required for post-replication repair of damaged DNA. Similar to its yeast counterpart, this protein is able to interact with the human homolog of yeast Rad6 protein through a conserved ring-finger motif. Mutation of this motif results in defective replication of UV-damaged DNA and hypersensitivity to multiple mutagens. [provided by RefSeq]
Sequence: MDSLAESRWPPGLAVMKTIDDLLRCGICFEYFNIAMIIPQCSHNYCSLCIRKFLSYKTQCPTCCVTVTEPDLKNNRILDELVKSLNFARNHLLQFALESPAKSPASSSSKNLAVKVYTPVASRQSLKQGSRLMDNFLIREMSGSTSELLIKENKSKFSPQKEASPAAKTKETRSVEEIAPDPSEAKRPEPPSTSTLKQVTKVDCPVCGVNIPESHINKHLDSCLSREEKKESLRSSVHKRKPLPKTVYNLLSDRDLKKKLKEHGLSIQGNKQQLIKRHQEFVHMYNAQCDALHPKSAAEIVQEIENIEKTRMRLEASKLNESVMVFTKDQTEKEIDEIHSKYRKKHKSEFQLLVDQARKGYKKIAGMSQKTVTITKEDESTEKLSSVCMGQEDNMTSVTNHFSQSKLDSPEELEPDREEDSSSCIDIQEVLSSSESDSCNSSSSDIIRDLLEEEEAWEASHKNDLQDTEISPRQNRRTRAAESAEIEPRNKRNRNEspecificaciones
NP_064550.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RNF73 | |
RAD18 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56852 | |
RAD18 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDSLAESRWPPGLAVMKTIDDLLRCGICFEYFNIAMIIPQCSHNYCSLCIRKFLSYKTQCPTCCVTVTEPDLKNNRILDELVKSLNFARNHLLQFALESPAKSPASSSSKNLAVKVYTPVASRQSLKQGSRLMDNFLIREMSGSTSELLIKENKSKFSPQKEASPAAKTKETRSVEEIAPDPSEAKRPEPPSTSTLKQVTKVDCPVCGVNIPESHINKHLDSCLSREEKKESLRSSVHKRKPLPKTVYNLLSDRDLKKKLKEHGLSIQGNKQQLIKRHQEFVHMYNAQCDALHPKSAAEIVQEIENIEKTRMRLEASKLNESVMVFTKDQTEKEIDEIHSKYRKKHKSEFQLLVDQARKGYKKIAGMSQKTVTITKEDESTEKLSSVCMGQEDNMTSVTNHFSQSKLDSPEELEPDREEDSSSCIDIQEVLSSSESDSCNSSSSDIIRDLLEEEEAWEASHKNDLQDTEISPRQNRRTRAAESAEIEPRNKRNRN | |
RUO | |
RAD18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |