missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RAB42 Full-length ORF (NP_689517.1, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00115273-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MATQGPDKVIFLLVGHKSDLQSTRCVSAQEAEELAASLGMAFVETSVKNNCNVDLAFDTLADAIQQALQQGDIKLEEGWGGVRLIHKTQIPRSPSRKQHSGPCQCEspecificaciones
NP_689517.1 | |
Liquid | |
115273 | |
RAB42 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATQGPDKVIFLLVGHKSDLQSTRCVSAQEAEELAASLGMAFVETSVKNNCNVDLAFDTLADAIQQALQQGDIKLEEGWGGVRLIHKTQIPRSPSRKQHSGPCQC | |
RUO | |
RAB42 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC45806/RP4-669K10.6 | |
RAB42 | |
Yes | |
wheat germ expression system |