Learn More
Abnova™ Human PTPRO Full-length ORF (AAH35960, 1 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005800-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a receptor-type protein tyrosine phosphatase containing a single intracellular catalytic domain with a characteristic signature motif. The gene product, which has a transmembrane domain, is an integral membrane protein. Several alternatively spliced transcript variants, some of which encode different isoforms of the protein, have been described. These variants exhibit tissue-specific expression. [provided by RefSeq]
Sequence: MGHLPTGIHGARRLLPLLWLFVLFKNATAFHVTVQDDNNIVVSLEASDVISPASVYVVKITGESKNYFFEFEEFNSTLPPPVIFKASYHGLYYIITLVVVNGNVVTKPSRSITVLTKPLPVTSVSIYDYKPSPETGVLFEIHYPEKYNVFTRVNISYWEGKDFRTMLYKDFFKGKTVFNHWLPGMCYSNITFQLVSEATFNKSTLVEYSGVSHEPKQHRTAPYPPQNISVRIVNLNKNNWEEQSGNFPEESFMRSQDTIGKEKLFHFTEETPEIPSGNISSGWPDFNSSDYETTSQPYWWDSASAAPESEDEFVSVLPMEYENNSTLSETEKSTSGSFSFFPVQMILTWLPPKPPTAFDGFHIHIEREENFTEYLMVDEEAHEFVAELKEPGKYKLSVTTFSSSGSCETRKSQSAKSLSFYISPSGEWIEELTEKPQHVSVHVLSSTTALMSWTSSQENYNSTIVSVVSLTCQKQKESQRLEKQYCTQVNSSKPIIENLVPGAQYQVVIYLRKGPLIGPPSDPVTFAIVPTGIKDLMLYPLGPTAVVLSWTRPYLGVFRKYVVEMFYFNPATMTSEWTTYYEIAATVSLTASVVIFPEspecificaciones
AAH35960 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.41kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLEPP1/PTP-U2/PTPU2 | |
PTPRO | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5800 | |
PTPRO (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGHLPTGIHGARRLLPLLWLFVLFKNATAFHVTVQDDNNIVVSLEASDVISPASVYVVKITGESKNYFFEFEEFNSTLPPPVIFKASYHGLYYIITLVVVNGNVVTKPSRSITVLTKPLPVTSVSIYDYKPSPETGVLFEIHYPEKYNVFTRVNISYWEGKDFRTMLYKDFFKGKTVFNHWLPGMCYSNITFQLVSEATFNKSTLVEYSGVSHEPKQHRTAPYPPQNISVRIVNLNKNNWEEQSGNFPEESFMRSQDTIGKEKLFHFTEETPEIPSGNISSGWPDFNSSDYETTSQPYWWDSASAAPESEDEFVSVLPMEYENNSTLSETEKSTSGSFSFFPVQMILTWLPPKPPTAFDGFHIHIEREENFTEYLMVDEEAHEFVAELKEPGKYKLSVTTFSSSGSCETRKSQSAKSLSFYISPSGEWIEELTEKPQHVSVHVLSSTTALMSWTSSQENYNSTIVSVVSLTCQKQKESQRLEKQYCTQVNSSKPIIENLVPGAQYQVVIYLRKGPLIGPPSDPVTFAIVPTGIKDLMLYPLGPTAVVLSWTRPYLGVFRKYVVEMFYFNPATMTSEWTTYYEIAATVSLTASVVIFP | |
RUO | |
PTPRO | |
Wheat Germ (in vitro) | |
GST | |
Liquid |