Learn More
Abnova™ Human PTPDC1 Full-length ORF (AAH67120.1, 1 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00138639-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene contains a characteristic motif of protein tyrosine phosphatases (PTPs). PTPs regulate activities of phosphoproteins through dephosphorylation. They are signaling molecules involved in the regulation of a wide variety of biological processes. The specific function of this protein has not yet been determined. Alternatively spliced transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: MAAGVLPQNEQPYSTLVNNSECVANMKGNLERPTPKYTKVGERLRHVIPGHMACSMACGGRACKYENPARWSEQEQAIKGVYSSWVTDNILAMARPSSELLEKYHIIDQFLSHGIKTIINLQRPGEHASCGNPLEQESGFTYLPEAFMEAGIYFYNFGWKDYGVASLTTILDMVKVMTFALQEGKVAIHCHAGLGRTGVLIACYLVFATRMTADQAIIFVRAKRPNSIQTRGQLLCVREFTQFLTPLRNIFSCCDPKAHAVTLPQYLIRQRHLLHGYEARLLKHVPKIIHLVCKLLLDLAENRPVMMKDVSEGPGLSAEIEKTMSEMVTMQLDKELLRHDSDFSNPPNPTAVAADFDNRGMIFSNEQQFDPLWKRRNVECLQPLTHLKRRLSYSDSDLKRAENLLEQGETPQTVPAQILVGHKPRQQKLISHCYIPQSPEPDLHKEALVRSTLSFWSQSKFGGLEGLKDNGSPIFHGRIIPKEAQQSGAFSADVSGSHSPGEPVSPSFANVHKDPNPAHQQVSHCQCKTHGVGSPGSVRQNSRTPRSPLDCGSSPKAQFLVEHETQDSKDLSEAASHSALQSELSAEARRILAAKALANLNESVEKEELKRKVEMWQKELNSRDGAWERICGERDPFILCSLMWSWVEQLKEPVITKEDVDMLVDRRADAAEALFLLEKGQHQTILCVLHCIVNLQTIPVDVEEAFLAHAIKAFTKVNFDSENGPTVYNTLKKIFKHTLEEKRKMTKDGPKPGLEspecificaciones
AAH67120.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ42922/PTP9Q22 | |
PTPDC1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
138639 | |
PTPDC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAGVLPQNEQPYSTLVNNSECVANMKGNLERPTPKYTKVGERLRHVIPGHMACSMACGGRACKYENPARWSEQEQAIKGVYSSWVTDNILAMARPSSELLEKYHIIDQFLSHGIKTIINLQRPGEHASCGNPLEQESGFTYLPEAFMEAGIYFYNFGWKDYGVASLTTILDMVKVMTFALQEGKVAIHCHAGLGRTGVLIACYLVFATRMTADQAIIFVRAKRPNSIQTRGQLLCVREFTQFLTPLRNIFSCCDPKAHAVTLPQYLIRQRHLLHGYEARLLKHVPKIIHLVCKLLLDLAENRPVMMKDVSEGPGLSAEIEKTMSEMVTMQLDKELLRHDSDFSNPPNPTAVAADFDNRGMIFSNEQQFDPLWKRRNVECLQPLTHLKRRLSYSDSDLKRAENLLEQGETPQTVPAQILVGHKPRQQKLISHCYIPQSPEPDLHKEALVRSTLSFWSQSKFGGLEGLKDNGSPIFHGRIIPKEAQQSGAFSADVSGSHSPGEPVSPSFANVHKDPNPAHQQVSHCQCKTHGVGSPGSVRQNSRTPRSPLDCGSSPKAQFLVEHETQDSKDLSEAASHSALQSELSAEARRILAAKALANLNESVEKEELKRKVEMWQKELNSRDGAWERICGERDPFILCSLMWSWVEQLKEPVITKEDVDMLVDRRADAAEALFLLEKGQHQTILCVLHCIVNLQTIPVDVEEAFLAHAIKAFTKVNFDSENGPTVYNTLKKIFKHTLEEKRKMTKDGPKPGL | |
RUO | |
PTPDC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |