Learn More
Abnova™ Human PSCD3 Full-length ORF (NP_004218.1, 1 a.a. - 399 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009265-P02.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the PSCD (pleckstrin homology, Sec7 and coiled-coil domains) family. PSCD family members have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. This encoded protein is involved in the control of Golgi structure and function, and it may have a physiological role in regulating ADP-ribosylation factor protein 6 (ARF) functions, in addition to acting on ARF1. [provided by RefSeq]
Sequence: MDEDGGGEGGGVPEDLSLEEREELLDIRRRKKELIDDIERLKYEIAEVMTEIDNLTSVEESKTTQRNKQIAMGRKKFNMDPKKGIQFLIENDLLQSSPEDVAQFLYKGEGLNKTVIGDYLGERDEFNIKVLQAFVELHEFADLNLVQALRQFLWSFRLPGEAQKIDRMMEAFASRYCLCNPGVFQSTDTCYVLSFAIIMLNTSLHNHNVRDKPTAERFIAMNRGINEGGDLPEELLRNLYESIKNEPFKIPEDDGNDLTHTFFNPDREGWLLKLGGRVKTWKRRWFILTDNCLYYFEYTTDKEPRGIIPLENLSIREVEDPRKPNCFELYNPSHKGQVIKACKTEADGRVVEGNHVVYRISAPSPEEKEEWMKSIKASISRDPFYDMLATRKRRIANKKEspecificaciones
NP_004218.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARNO3/GRP1/PSCD3 | |
CYTH3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9265 | |
PSCD3 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDEDGGGEGGGVPEDLSLEEREELLDIRRRKKELIDDIERLKYEIAEVMTEIDNLTSVEESKTTQRNKQIAMGRKKFNMDPKKGIQFLIENDLLQSSPEDVAQFLYKGEGLNKTVIGDYLGERDEFNIKVLQAFVELHEFADLNLVQALRQFLWSFRLPGEAQKIDRMMEAFASRYCLCNPGVFQSTDTCYVLSFAIIMLNTSLHNHNVRDKPTAERFIAMNRGINEGGDLPEELLRNLYESIKNEPFKIPEDDGNDLTHTFFNPDREGWLLKLGGRVKTWKRRWFILTDNCLYYFEYTTDKEPRGIIPLENLSIREVEDPRKPNCFELYNPSHKGQVIKACKTEADGRVVEGNHVVYRISAPSPEEKEEWMKSIKASISRDPFYDMLATRKRRIANKK | |
RUO | |
CYTH3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |