Learn More
Abnova™ Human PRV1 Partial ORF (AAH29167, 21 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057126-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
NB1, a glycosyl-phosphatidylinositol (GPI)-linked N-glycosylated cell surface glycoprotein, was first described in a case of neonatal alloimmune neutropenia (Lalezari et al., 1971 [PubMed 5552408]).[supplied by OMIM]
Sequence: ALLCQFGTVQHVWKVSDLPRQWTPKNTSCDSGLGCQDTLMLIESGPQVSLVLSKGCTEAKDQEPRVTEHRMGPGLSLISYTFVCRQEDFCNNLVNSLPLWEspecificaciones
AAH29167 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ALLCQFGTVQHVWKVSDLPRQWTPKNTSCDSGLGCQDTLMLIESGPQVSLVLSKGCTEAKDQEPRVTEHRMGPGLSLISYTFVCRQEDFCNNLVNSLPLW | |
RUO | |
CD177 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57126 | |
PRV1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HNA2A/NB1/PRV1 | |
CD177 | |
Recombinant | |
wheat germ expression system |