missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRPSAP2 Full-length ORF (NP_002758.1, 1 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005636-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16169691

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS. [provided by RefSeq]

Sequence: MFCVTPPELETKMNITKGGLVLFSANSNSSCMELSKKIAERLGVEMGKVQVYQEPNRETRVQIQESVRGKDVFIIQTVSKDVNTTIMELLIMVYACKTSCAKSIIGVIPYFPYSKQCKMRKRGSIVSKLLASMMCKAGLTHLITMDLHQKEIQGFFNIPVDNLRASPFLLQYIQEEIPDYRNAVIVAKSPASAKRAQSFAERLRLGIAVIHGEAQDAESDLVDGRHSPPMVRSVAAIHPSLEIPMLIPKEKPPITVVGDVGGRIAIIVDDIIDDVDSFLAAAETLKERGAYKIFVMATHGLLSSDAPRRIEESAIDEVVVTNTIPHEVQKLQCPKIKTVDISMILSEAIRRIHNGESMSYLFRNIGLDD
Especificaciones

Especificaciones

NP_002758.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC117304/MGC126719/MGC126721/PAP41
PRPSAP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5636
PRPSAP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MFCVTPPELETKMNITKGGLVLFSANSNSSCMELSKKIAERLGVEMGKVQVYQEPNRETRVQIQESVRGKDVFIIQTVSKDVNTTIMELLIMVYACKTSCAKSIIGVIPYFPYSKQCKMRKRGSIVSKLLASMMCKAGLTHLITMDLHQKEIQGFFNIPVDNLRASPFLLQYIQEEIPDYRNAVIVAKSPASAKRAQSFAERLRLGIAVIHGEAQDAESDLVDGRHSPPMVRSVAAIHPSLEIPMLIPKEKPPITVVGDVGGRIAIIVDDIIDDVDSFLAAAETLKERGAYKIFVMATHGLLSSDAPRRIEESAIDEVVVTNTIPHEVQKLQCPKIKTVDISMILSEAIRRIHNGESMSYLFRNIGLDD
RUO
PRPSAP2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales