Learn More
Abnova™ Human PRDM5 Full-length ORF (AAH66942.1, 1 a.a. - 111 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00011107-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. [provided by RefSeq]
Sequence: MLGMYVPDRFSLKSSRVQDGMGLYTARRVRKGEKFGPFAGEKRMPEDLDENMDYRLMWEVRGSKGEVLYILDATNPRHSNWLRFVHEAPSQEQKNLAAIQDKNLGPAEWRGEspecificaciones
AAH66942.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PFM2 | |
PRDM5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11107 | |
PRDM5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGMYVPDRFSLKSSRVQDGMGLYTARRVRKGEKFGPFAGEKRMPEDLDENMDYRLMWEVRGSKGEVLYILDATNPRHSNWLRFVHEAPSQEQKNLAAIQDKNLGPAEWRG | |
RUO | |
PRDM5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |