Learn More
Abnova™ Human POU3F2 Full-length ORF (AAH51699.2, 1 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005454-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
POU3F2 belongs to a large family of transcription factors that bind to the octameric DNA sequence ATGCAAAT. Most of these proteins share a highly homologous region, referred to as the POU domain, that occurs in several mammalian transcription factors, including the octamer-binding proteins Oct1 (POU2F1; MIM 164175) and Oct2 (POU2F2; MIM 164176) and the pituitary protein Pit1 (PIT1; MIM 173110). Class III POU genes are expressed predominantly in the central nervous system (CNS). It is likely that CNS-specific transcription factors such as these play an important role in mammalian neurogenesis by regulating their diverse patterns of gene expression (Schreiber et al., 1993 [PubMed 8441633]; Atanasoski et al., 1995 [PubMed 7601453]).[supplied by OMIM]
Sequence: MATAASNHYSLLTSSASIVHAEPPGGMQQGAGGYREAQSLVQGDYGALQSNGHPLSHAHQWITALSHGGGGGGGGGGGGGGGGGGGGGDGSPWSTSPLGQPDIKPSVVVQQGGRGDELHGPGALQQQHQQQQQQQQQQQQQQQQQQQQQRPPHLVHHAANHHPGPGAWRSAAAAAHLPPSMGASNGGLLYSQPSFTVNGMLGAGGQSAGLHHHGLRDAHDEPHHADHHPHPHSHPHQQPPPPPPPQGPPGHPGAHHDPHSDEDTPTSDDLEQFAKQFKQRRIKLGFTQADVGLALGTLYGNVFSQTTICRFEALQLSFKNMCKLKPLLNKWLEEADSSSGSPTSIDKIAAQGRKRKKRTSIEVSVKGALESHFLKCPKPSAQEITSLADSLQLEKEVVRVWFCNRRQKEKRMTPPGGTLPGAEDVYGGSRDTPPHHGVQTPVQEspecificaciones
AAH51699.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRN2/OCT7/OTF7/POUF3 | |
POU3F2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5454 | |
POU3F2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATAASNHYSLLTSSASIVHAEPPGGMQQGAGGYREAQSLVQGDYGALQSNGHPLSHAHQWITALSHGGGGGGGGGGGGGGGGGGGGGDGSPWSTSPLGQPDIKPSVVVQQGGRGDELHGPGALQQQHQQQQQQQQQQQQQQQQQQQQQRPPHLVHHAANHHPGPGAWRSAAAAAHLPPSMGASNGGLLYSQPSFTVNGMLGAGGQSAGLHHHGLRDAHDEPHHADHHPHPHSHPHQQPPPPPPPQGPPGHPGAHHDPHSDEDTPTSDDLEQFAKQFKQRRIKLGFTQADVGLALGTLYGNVFSQTTICRFEALQLSFKNMCKLKPLLNKWLEEADSSSGSPTSIDKIAAQGRKRKKRTSIEVSVKGALESHFLKCPKPSAQEITSLADSLQLEKEVVRVWFCNRRQKEKRMTPPGGTLPGAEDVYGGSRDTPPHHGVQTPVQ | |
RUO | |
POU3F2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |