Learn More
Abnova™ Human POLD3 Partial ORF (NP_006582, 357 a.a. - 466 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010714-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The DNA polymerase delta complex is involved in DNA replication and repair, and it consists of the proliferating cell nuclear antigen (PCNA; MIM 176740), the multisubunit replication factor C (see MIM 102579), and the 4 subunit polymerase complex: POLD1 (MIM 174761), POLD2 (MIM 600815), POLD3, and POLD4 (MIM 611525) (Liu and Warbrick, 2006 [PubMed 16934752]).[supplied by OMIM]
Sequence: PPLEPVPKTEPEPPSVKSSSGENKRKRKRVLKSKTYLDGEGCIVTEKVYESESCTDSEEELNMKTSSVHRPPAMTVKKEPREERKGPKKGTAALGKANRQVSITGFFQRKEspecificaciones
NP_006582 | |
Liquid | |
10714 | |
POLD3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0039/MGC119642/MGC119643/P66/P68 | |
POLD3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PPLEPVPKTEPEPPSVKSSSGENKRKRKRVLKSKTYLDGEGCIVTEKVYESESCTDSEEELNMKTSSVHRPPAMTVKKEPREERKGPKKGTAALGKANRQVSITGFFQRK | |
RUO | |
POLD3 | |
Wheat Germ (in vitro) | |
GST |