Learn More
Abnova™ Human PNPLA3 Full-length ORF (AAH65195.1, 1 a.a. - 481 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080339-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes. [provided by RefSeq]
Sequence: MYDAERGWSLSFAGCGFLGFYHVGATRCLSEHAPHLLRDARMLFGASAGALHCVGVLSGIPLEQTLQVLSDLVRKARSRNIGIFHPSFNLSKFLRQGLGKCLPANVHQLISGKIGISLTRVSDGENVLVSDFRSKDEVVDALVCSCFMPFYSGLIPPSFRGVRYVDGGVSDNVPFIDAKTTITVSPFYGEYDICPKVKSTNFLHVDITKLSLRLCTGNLYLLSRAFVPPDLKVLGEICLRGYLDAFRFLEEKGICNRPQPGLKSSSEGMDPEVAMPSWANMSLDSSPESAALAVRLEGDELLDHLRLSILPWDESILDTLSPRLATALSEEMKDKGGYMSKICNLLPIRIMSYVMLPCTLPVESAIAIVQRLVTWLPDMPDDVLWLQWVTSQVFTRVLMCLLPASRSQMPVSSQQASPCTPEQDWPCWTPCSPEGCPAETKAEATPRSILRSSLNFFLGNKVPAGAEGLSTFPSFSLEKSLEspecificaciones
AAH65195.1 | |
Liquid | |
80339 | |
PNPLA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MYDAERGWSLSFAGCGFLGFYHVGATRCLSEHAPHLLRDARMLFGASAGALHCVGVLSGIPLEQTLQVLSDLVRKARSRNIGIFHPSFNLSKFLRQGLGKCLPANVHQLISGKIGISLTRVSDGENVLVSDFRSKDEVVDALVCSCFMPFYSGLIPPSFRGVRYVDGGVSDNVPFIDAKTTITVSPFYGEYDICPKVKSTNFLHVDITKLSLRLCTGNLYLLSRAFVPPDLKVLGEICLRGYLDAFRFLEEKGICNRPQPGLKSSSEGMDPEVAMPSWANMSLDSSPESAALAVRLEGDELLDHLRLSILPWDESILDTLSPRLATALSEEMKDKGGYMSKICNLLPIRIMSYVMLPCTLPVESAIAIVQRLVTWLPDMPDDVLWLQWVTSQVFTRVLMCLLPASRSQMPVSSQQASPCTPEQDWPCWTPCSPEGCPAETKAEATPRSILRSSLNFFLGNKVPAGAEGLSTFPSFSLEKSL | |
RUO | |
PNPLA3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADPN/C22orf20/iPLA(2)epsilon | |
PNPLA3 | |
Yes | |
wheat germ expression system |