missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PMS2 Full-length ORF (NP_000526.1, 1 a.a. - 862 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005395-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16169251

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is one of the PMS2 gene family members found in clusters on chromosome 7. The product of this gene is involved in DNA mismatch repair. It forms a heterodimer with MLH1 and this complex interacts with other complexes bound to mismatched bases. Mutations in this gene are associated with hereditary nonpolyposis colorectal cancer, Turcot syndrome, and are a cause of supratentorial primitive neuroectodermal tumors. Alternatively spliced transcript variants have been observed for this gene. [provided by RefSeq]

Sequence: MERAESSSTEPAKAIKPIDRKSVHQICSGQVVLSLSTAVKELVENSLDAGATNIDLKLKDYGVDLIEVSDNGCGVEEENFEGLTLKHHTSKIQEFADLTQVETFGFRGEALSSLCALSDVTISTCHASAKVGTRLMFDHNGKIIQKTPYPRPRGTTVSVQQLFSTLPVRHKEFQRNIKKEYAKMVQVLHAYCIISAGIRVSCTNQLGQGKRQPVVCTGGSPSIKENIGSVFGQKQLQSLIPFVQLPPSDSVCEEYGLSCSDALHNLFYISGFISQCTHGVGRSSTDRQFFFINRRPCDPAKVCRLVNEVYHMYNRHQYPFVVLNISVDSECVDINVTPDKRQILLQEEKLLLAVLKTSLIGMFDSDVNKLNVSQQPLLDVEGNLIKMHAADLEKPMVEKQDQSPSLRTGEEKKDVSISRLREAFSLRHTTENKPHSPKTPEPRRSPLGQKRGMLSSSTSGAISDKGVLRPQKEAVSSSHGPSDPTDRAEVEKDSGHGSTSVDSEGFSIPDTGSHCSSEYAASSPGDRGSQEHVDSQEKAPETDDSFSDVDCHSNQEDTGCKFRVLPQPTNLATPNTKRFKKEEILSSSDICQKLVNTQDMSASQVDVAVKINKKVVPLDFSMSSLAKRIKQLHHEAQQSEGEQNYRKFRAKICPGENQAAEDELRKEISKTMFAEMEIIGQFNLGFIITKLNEDIFIVDQHATDEKYNFEMLQQHTVLQGQRLIAPQTLNLTAVNEAVLIENLEIFRKNGFDFVIDENAPVTERAKLISLPTSKNWTFGPQDVDELIFMLSDSPGVMCRPSRVKQMFASRACRKSVMIGTALNTSEMKKLITHMGEMDHPWNCPHGRPTMRHIANLGVISQN
Especificaciones

Especificaciones

NP_000526.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
122.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HNPCC4/PMS2CL/PMSL2
PMS2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5395
PMS2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERAESSSTEPAKAIKPIDRKSVHQICSGQVVLSLSTAVKELVENSLDAGATNIDLKLKDYGVDLIEVSDNGCGVEEENFEGLTLKHHTSKIQEFADLTQVETFGFRGEALSSLCALSDVTISTCHASAKVGTRLMFDHNGKIIQKTPYPRPRGTTVSVQQLFSTLPVRHKEFQRNIKKEYAKMVQVLHAYCIISAGIRVSCTNQLGQGKRQPVVCTGGSPSIKENIGSVFGQKQLQSLIPFVQLPPSDSVCEEYGLSCSDALHNLFYISGFISQCTHGVGRSSTDRQFFFINRRPCDPAKVCRLVNEVYHMYNRHQYPFVVLNISVDSECVDINVTPDKRQILLQEEKLLLAVLKTSLIGMFDSDVNKLNVSQQPLLDVEGNLIKMHAADLEKPMVEKQDQSPSLRTGEEKKDVSISRLREAFSLRHTTENKPHSPKTPEPRRSPLGQKRGMLSSSTSGAISDKGVLRPQKEAVSSSHGPSDPTDRAEVEKDSGHGSTSVDSEGFSIPDTGSHCSSEYAASSPGDRGSQEHVDSQEKAPETDDSFSDVDCHSNQEDTGCKFRVLPQPTNLATPNTKRFKKEEILSSSDICQKLVNTQDMSASQVDVAVKINKKVVPLDFSMSSLAKRIKQLHHEAQQSEGEQNYRKFRAKICPGENQAAEDELRKEISKTMFAEMEIIGQFNLGFIITKLNEDIFIVDQHATDEKYNFEMLQQHTVLQGQRLIAPQTLNLTAVNEAVLIENLEIFRKNGFDFVIDENAPVTERAKLISLPTSKNWTFGPQDVDELIFMLSDSPGVMCRPSRVKQMFASRACRKSVMIGTALNTSEMKKLITHMGEMDHPWNCPHGRPTMRHIANLGVISQN
RUO
PMS2
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales