missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PLXNA3 Partial ORF (NP_059984, 23 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055558-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: FRAFVVTDTTLTHLAVHRVTGEVFVGAVNRVFKLAPNLTELRAHVTGPVEDNARCYPPPSMRVCAHRLAPVDNINKLLLIDYAARRLVACGSIWQGICQFLRLDDLEspecificaciones
NP_059984 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.4kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FRAFVVTDTTLTHLAVHRVTGEVFVGAVNRVFKLAPNLTELRAHVTGPVEDNARCYPPPSMRVCAHRLAPVDNINKLLLIDYAARRLVACGSIWQGICQFLRLDDL | |
RUO | |
PLXNA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55558 | |
PLXNA3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
6.3/HSSEXGENE/PLEXIN-A3/PLXN3/PLXN4/SEX/XAP-6 | |
PLXNA3 | |
Recombinant | |
wheat germ expression system |