Learn More
Abnova™ Human PLP1 Partial ORF (NP_000524, 177 a.a. - 232 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005354-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene. [provided by RefSeq]
Sequence: YFNTWTTCQSIAFPSKTSASIGSLCADARMYGVLPWNAFPGKVCGSNLLSICKTAEEspecificaciones
NP_000524 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.9kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YFNTWTTCQSIAFPSKTSASIGSLCADARMYGVLPWNAFPGKVCGSNLLSICKTAE | |
RUO | |
PLP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5354 | |
PLP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HLD1/MMPL/PLP/PLP/DM20/PMD/SPG2 | |
PLP1 | |
Recombinant | |
wheat germ expression system |