missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PHOSPHO1 Partial ORF (NP_848595, 168 a.a. - 266 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00162466-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: CARCPANMCKHKVLSDYLRERAHDGVHFERLFYVGDGANDFCPMGLLAGGDVAFPRRGYPMHRLIQEAQKAEPSSFRASVVPWETAADVRLHLQQVLKSEspecificaciones
NP_848595 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CARCPANMCKHKVLSDYLRERAHDGVHFERLFYVGDGANDFCPMGLLAGGDVAFPRRGYPMHRLIQEAQKAEPSSFRASVVPWETAADVRLHLQQVLKS | |
RUO | |
PHOSPHO1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
162466 | |
PHOSPHO1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PHOSPHO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |