Learn More
Abnova™ Human PDCD7 Partial ORF (AAH16992, 47 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010081-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with sequence similarity to a mouse protein originally identified in embryonic stem cells. In mouse T-cell lines, this protein appears to be related to glucocorticoid- and staurine-induced apoptotic pathways, and to be linked to ceramide-mediated signalling. These observations suggest that this gene product is involved in specific apoptotic processes in T-cells. [provided by RefSeq]
Sequence: KRELEKKQRKEKEKILLQKREIESKLFGDPDEFPLAHLLEPFRQYYLQAEHSLPALIQIRHDWDQYLVPSDHPKGNFVPQGWVLPPLPSNDIWATAVKLHEspecificaciones
AAH16992 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KRELEKKQRKEKEKILLQKREIESKLFGDPDEFPLAHLLEPFRQYYLQAEHSLPALIQIRHDWDQYLVPSDHPKGNFVPQGWVLPPLPSNDIWATAVKLH | |
RUO | |
PDCD7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10081 | |
PDCD7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ES18/HES18/MGC22015 | |
PDCD7 | |
Recombinant | |
wheat germ expression system |