Learn More
Abnova™ Human PCGF1 Full-length ORF (AAH04952.1, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084759-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
PCGF1 is a mammalian homolog of the Drosophila polycomb group genes, which act as transcriptional repressors to regulate anterior-posterior patterning in early embryonic development (Nunes et al., 2001 [PubMed 11287196]). See also PCGF2 (MIM 600346).[supplied by OMIM]
Sequence: MRLRNQLQSVYKMDPLRNEEEVRVKIKDLNEHIVCCLCAGYFVDATTITECLHTFCKSCIVKYLQTSKYCPMCNIKIHETQPLLNLKLDRVMQDIVYKLVPGLQDSEEKRIREFYQSRGLDRVTQPTGEEPALSNLGLPFSSFDHSKAHYYRYDEQLNLCLERLSSGKDKNKSVLQNKYVRCSVRAEVRHLRRVLCHRLMLNPQHVQLLFDNEVLPDHMTMKQIWLSRWFGKPSPLLLQYSVKEKRREspecificaciones
AAH04952.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
2010002K04Rik/FLJ43754/MGC10882/NSPC1/RNF3A-2/RNF68 | |
PCGF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84759 | |
PCGF1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRLRNQLQSVYKMDPLRNEEEVRVKIKDLNEHIVCCLCAGYFVDATTITECLHTFCKSCIVKYLQTSKYCPMCNIKIHETQPLLNLKLDRVMQDIVYKLVPGLQDSEEKRIREFYQSRGLDRVTQPTGEEPALSNLGLPFSSFDHSKAHYYRYDEQLNLCLERLSSGKDKNKSVLQNKYVRCSVRAEVRHLRRVLCHRLMLNPQHVQLLFDNEVLPDHMTMKQIWLSRWFGKPSPLLLQYSVKEKRR | |
RUO | |
PCGF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |