Learn More
Abnova™ Human PCGF1 Full-length ORF (AAH04952.1, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00084759-P01.10ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16166063
  • 335.00€ / 10 microgramos
Fecha estimada de envío: 13-06-2024
para ver el stock
Descripción
Descripción
PCGF1 is a mammalian homolog of the Drosophila polycomb group genes, which act as transcriptional repressors to regulate anterior-posterior patterning in early embryonic development (Nunes et al., 2001 [PubMed 11287196]). See also PCGF2 (MIM 600346).[supplied by OMIM]
Sequence: MRLRNQLQSVYKMDPLRNEEEVRVKIKDLNEHIVCCLCAGYFVDATTITECLHTFCKSCIVKYLQTSKYCPMCNIKIHETQPLLNLKLDRVMQDIVYKLVPGLQDSEEKRIREFYQSRGLDRVTQPTGEEPALSNLGLPFSSFDHSKAHYYRYDEQLNLCLERLSSGKDKNKSVLQNKYVRCSVRAEVRHLRRVLCHRLMLNPQHVQLLFDNEVLPDHMTMKQIWLSRWFGKPSPLLLQYSVKEKRR
Especificaciones
Especificaciones
AAH04952.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
2010002K04Rik/FLJ43754/MGC10882/NSPC1/RNF3A-2/RNF68
PCGF1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84759
PCGF1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRLRNQLQSVYKMDPLRNEEEVRVKIKDLNEHIVCCLCAGYFVDATTITECLHTFCKSCIVKYLQTSKYCPMCNIKIHETQPLLNLKLDRVMQDIVYKLVPGLQDSEEKRIREFYQSRGLDRVTQPTGEEPALSNLGLPFSSFDHSKAHYYRYDEQLNLCLERLSSGKDKNKSVLQNKYVRCSVRAEVRHLRRVLCHRLMLNPQHVQLLFDNEVLPDHMTMKQIWLSRWFGKPSPLLLQYSVKEKRR
RUO
PCGF1