Learn More
Abnova™ Human PCDHGB6 Full-length ORF (NP_115271.1, 1 a.a. - 820 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056100-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]
Sequence: MGGSCAQRRRAGPRQVLFPLLLPLFYPTLSEPIRYSIPEELAKGSVVGNLAKDLGLSVLDVSARKLRVSAEKLHFSVDAESGDLLVKNRIDREQICKERRRCELQLEAVVENPLNIFHVIVVIEDVNDHAPQFDKKEIHLEIFESASAGTRLSLDPATDPDININSIKDYKINSNPYFSLMVRVNSDGGKYPELSLEKLLDREEQRSHSLILTALDGGDPPRSATAHIEISVKDTNDNPPVFSRDEYRISLSENLPPGSPVLQVTATDQDEGVNAEINYYFRSTAQSTKHMFSLDEKTGMIKNNQSFDFEDVERYTMEVEAKDGGGLSTQCKVIIEILDENDNSPEIIITSLSDQILENSPPGMVVALFKTRDLDFGGNGEVRCNIETDIPFKIYSSSNNYYKLVTDGALDREQTPEYNVTIVATDRGKPPLSSSRSITLYVADINDNAPVFDQTSYVVHVAENNPPGASIAQVSASDPDLGLNGHISYSIVASDLEPLAVSSYVSVSAQSGVVFAQRAFDHEQLRAFALTLQARDHGSPTLSANVSLRVLVGDRNDNAPRVLYPALGPDGSAFFDMVPRSAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLVFADNLQEILPDLSDRPVLSDPQAELQFYLVVALALISVLFLLAVILAIALRLRRSLSPATWDCFHPGLCVKSGPVVPPNYSEGTLPYSYNLCIAHTGTKEFNFLKCSVPLHSNEDMVCSVSPGALIPPHGGEDLTSHPETLTSVSFSFLCVIYLIVYEspecificaciones
NP_115271.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
116.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCDH-GAMMA-B6 | |
PCDHGB6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56100 | |
PCDHGB6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGGSCAQRRRAGPRQVLFPLLLPLFYPTLSEPIRYSIPEELAKGSVVGNLAKDLGLSVLDVSARKLRVSAEKLHFSVDAESGDLLVKNRIDREQICKERRRCELQLEAVVENPLNIFHVIVVIEDVNDHAPQFDKKEIHLEIFESASAGTRLSLDPATDPDININSIKDYKINSNPYFSLMVRVNSDGGKYPELSLEKLLDREEQRSHSLILTALDGGDPPRSATAHIEISVKDTNDNPPVFSRDEYRISLSENLPPGSPVLQVTATDQDEGVNAEINYYFRSTAQSTKHMFSLDEKTGMIKNNQSFDFEDVERYTMEVEAKDGGGLSTQCKVIIEILDENDNSPEIIITSLSDQILENSPPGMVVALFKTRDLDFGGNGEVRCNIETDIPFKIYSSSNNYYKLVTDGALDREQTPEYNVTIVATDRGKPPLSSSRSITLYVADINDNAPVFDQTSYVVHVAENNPPGASIAQVSASDPDLGLNGHISYSIVASDLEPLAVSSYVSVSAQSGVVFAQRAFDHEQLRAFALTLQARDHGSPTLSANVSLRVLVGDRNDNAPRVLYPALGPDGSAFFDMVPRSAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLVFADNLQEILPDLSDRPVLSDPQAELQFYLVVALALISVLFLLAVILAIALRLRRSLSPATWDCFHPGLCVKSGPVVPPNYSEGTLPYSYNLCIAHTGTKEFNFLKCSVPLHSNEDMVCSVSPGALIPPHGGEDLTSHPETLTSVSFSFLCVIYLIVY | |
RUO | |
PCDHGB6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |