Learn More
Abnova™ Human PCDH21 Full-length ORF (AAH38799.1, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00092211-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Beschreibung
This gene is a member of the cadherin superfamily of calcium-dependent cell-cell adhesion molecules. The encoded protein has a signal peptide, six cadherin repeat domains and a unique cytoplasmic region. This non-classical cadherin appears to be exclusively expressed in the mitral and tufted cells in the main and accessory olfactory bulbs of the brain, suggesting a possible role in the formation and maintenance of neuronal networks. [provided by RefSeq]
Sequence: MRRCRWAALALGLLRLCLAQANFAPHFFDNGVGSTNGNMALFSLPEDTPVGSHVYTLNGTDPEGDPISYHISFDPSTRSVFSVDPTFGNITLVEELDREREDEIEAIISISDGLNLVAEKVVILVTDANDEAPRFIQEPYVALVPEDIPAGSIIFKVHAVDRDTGSGGSVTYFLQNLHSPFAVDRHSGVLRLQAGATLDYERSRTHYITVVAKDGGGRLHGADVVFSATTTVTVNVEDVQDMAPVFVGTPYYGYVYEDTLPGSEVLKVVAMDGDRGKPNRILYSLVNGNDGAFEINETSGAISITQSPAQLQREVYELHVQVTEMSPAGSPAAQATVPVTIRIVDLNNHPPTFYGESGPQNRFELSMNEHPPQGEILRGLKITVNDSDQGANAKFNLQLVGPRGIFRVVPQTVLNEAQVTIIVENSAAIDFEKSKVLTFKLLAVEVNTPEKFSSTADVVIQLLDTNDNVPKFDSLYYVARIPENAPGGSSVVAVTAVDPDTGPWGEVKYSTYGTGADLFLIHPSTGLIYTQPWASLDAEATARYNFYVKAEDMEGKYSVAEVFITLLDVNDHPPQFGKSVQKKTMVLGTPVKIEAIDEDAEEPNNLVDYSITHAEPANVFDINSHTGEIWLKNSIRSLDALHNITPGRDCLWSLEVQAKDRGSPSFSTTALLKIDITDAEVRRLRYMKNSNFPGTTKSVRKPKFKPKKPHSSQGLFLHPHCEIALFNLSNVNLYSRVFQGAAQASSpezifikation
AAH38799.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
107.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434A132/KIAA1775/PRCAD | |
PCDH21 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
92211 | |
PCDH21 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRCRWAALALGLLRLCLAQANFAPHFFDNGVGSTNGNMALFSLPEDTPVGSHVYTLNGTDPEGDPISYHISFDPSTRSVFSVDPTFGNITLVEELDREREDEIEAIISISDGLNLVAEKVVILVTDANDEAPRFIQEPYVALVPEDIPAGSIIFKVHAVDRDTGSGGSVTYFLQNLHSPFAVDRHSGVLRLQAGATLDYERSRTHYITVVAKDGGGRLHGADVVFSATTTVTVNVEDVQDMAPVFVGTPYYGYVYEDTLPGSEVLKVVAMDGDRGKPNRILYSLVNGNDGAFEINETSGAISITQSPAQLQREVYELHVQVTEMSPAGSPAAQATVPVTIRIVDLNNHPPTFYGESGPQNRFELSMNEHPPQGEILRGLKITVNDSDQGANAKFNLQLVGPRGIFRVVPQTVLNEAQVTIIVENSAAIDFEKSKVLTFKLLAVEVNTPEKFSSTADVVIQLLDTNDNVPKFDSLYYVARIPENAPGGSSVVAVTAVDPDTGPWGEVKYSTYGTGADLFLIHPSTGLIYTQPWASLDAEATARYNFYVKAEDMEGKYSVAEVFITLLDVNDHPPQFGKSVQKKTMVLGTPVKIEAIDEDAEEPNNLVDYSITHAEPANVFDINSHTGEIWLKNSIRSLDALHNITPGRDCLWSLEVQAKDRGSPSFSTTALLKIDITDAEVRRLRYMKNSNFPGTTKSVRKPKFKPKKPHSSQGLFLHPHCEIALFNLSNVNLYSRVFQGAAQAS | |
RUO | |
PCDH21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |