missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human P53AIP1 Partial ORF (NP_071395, 1 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00063970-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MGSSSEASFRSAQASCSGARRQGLGRGDQNLSVMPPNGRAQTHTPGWVSPCSENRDGLLPATAPGRLCSHRGADIPSFQTHQDPVTASGSSELHADCPQFRALDRAGNEspecificaciones
NP_071395 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGSSSEASFRSAQASCSGARRQGLGRGDQNLSVMPPNGRAQTHTPGWVSPCSENRDGLLPATAPGRLCSHRGADIPSFQTHQDPVTASGSSELHADCPQFRALDRAGN | |
RUO | |
P53AIP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
63970 | |
P53AIP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
P53AIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |